Learn More
Ace Glass, Inc. Combo mantle, 6 position, high temperature, fits 100 to 125mLround bottom flasks, max diameter 2.75 inches, 120v 140 watts per position, order code (-65) or (-75) power supply separately
SDP
Catalog No. 50977349
Encompass
Combo mantle, 6 position, high temperature, fits 100 to 125mLround bottom flasks, max diameter 2.75 inches, 120v 140 watts per position, order code (-65) or (-75) power supply separately
Catalog No. 50-977-349 Supplier Ace Glass, Inc. Supplier No. 1206109
Combo mantle, 6 position, high temperature, fits 100 to 125mLround bottom flasks, max diameter 2.75 inches, 120v 140 watts per position, order code (-65) or (-75) power supply separately