Learn More
Ace Glass, Inc. Combo mantle, 6 position, low temperature, fits 100 to 125mLround bottom flasks, max diameter 2.75 inches, 120v 85 watts per position, order code (-65) or (-75) power supply separately
SDP
Catalog No. 50977348
Encompass
Combo mantle, 6 position, low temperature, fits 100 to 125mLround bottom flasks, max diameter 2.75 inches, 120v 85 watts per position, order code (-65) or (-75) power supply separately
Catalog No. 50-977-348 Supplier Ace Glass, Inc. Supplier No. 1206105
Combo mantle, 6 position, low temperature, fits 100 to 125mLround bottom flasks, max diameter 2.75 inches, 120v 85 watts per position, order code (-65) or (-75) power supply separately