Learn More
Waters Corp Waters QC Trace Metal Standard, Selenium, 1000 mg/L, 125 mL
Catalog No. 50465854 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This standard is a Selenium QC Trace Metal Standard with a concentration of 1000 mg/L and a Nitric Acid matrix. It is designed for environmental analysis using Inductively Coupled Plasma-Optical Emission Spectrometry (ICP-OES), Inductively Coupled Plasma-Mass Spectrometry (ICP-MS), and Atomic Absorption (AA) methods.
  • Liquid formulation
  • Designed for environmental analysis
  • Compatible with ICP-OES, ICP-MS, and AA methods
Catalog No. 50-465-854 Supplier Waters Corp Supplier No. 031
Add to Cart
Add to Cart
This standard is a Selenium QC Trace Metal Standard with a concentration of 1000 mg/L and a Nitric Acid matrix. It is designed for environmental analysis using Inductively Coupled Plasma-Optical Emission Spectrometry (ICP-OES), Inductively Coupled Plasma-Mass Spectrometry (ICP-MS), and Atomic Absorption (AA) methods.
  • Liquid formulation
  • Designed for environmental analysis
  • Compatible with ICP-OES, ICP-MS, and AA methods