Learn More
Chemglass Life Sciences 125mL Sep Funnel, Old Style, 19/22
SDP
Catalog No. 50873283
Encompass_Preferred
125mL Separatory Funnel, Squibb, 19/22 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.
Catalog No. 50-873-283 Supplier Chemglass Life Sciences Supplier No. CG174351
125mL Separatory Funnel, Squibb, 19/22 Joint Size. Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.