Learn More
Chemglass Life Sciences 125mL Sep Funnel, Old Style, 14/20
SDP
Catalog No. 501531219
Encompass_Preferred
Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.
Catalog No. 50-153-1219 Supplier Chemglass Life Sciences Supplier No. CG174350
Add to Cart
Add to Cart
Squibb style separatory funnel with a PTFE stopcock. Not supplied with stopper. For stoppers see CG-3000. Stopcock bore on all sizes is 2mm.