Learn More
Waters Corp Waters Silica, Ion Calibration Standard, 1000 mg/L, 125 mL, Liquid
Catalog No. 50465872 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Silica, Ion Calibration Standard is a 1000 mg/L liquid formulation. It is designed for use in primary calibration or for preparing second source calibration check standards.
  • Can be used for primary calibration or to prepare second source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available).
  • Guaranteed stable for one year.
Catalog No. 50-465-872 Supplier Waters Corp Supplier No. 064
This Silica, Ion Calibration Standard is a 1000 mg/L liquid formulation. It is designed for use in primary calibration or for preparing second source calibration check standards.
  • Can be used for primary calibration or to prepare second source calibration check standards.
  • Traceable to NIST Standard Reference Materials (where available).
  • Guaranteed stable for one year.