Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Sodium Standard, 1000 mg/L, 125 mL

Supplier:  Waters Corp 034

Encompass_Preferred

This product is a QC Trace Metal Standard for Sodium, with a concentration of 1000 mg/L in an HNO3 matrix.

  • Use with ICP-OES, ICP-MS and AA methods.
  • One 15 mL screw-cap vial yields up to 2 L after dilution.
  • Liquid formulation.
  • Suitable for environmental applications.
  • Classified as a standard product type.
  • Contains hazardous material requiring special freight handling; additional charges may apply.

Catalog No. 50-465-857


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.