Learn More
Waters Corp 125 mL Sulphate
Supplier:  Waters Corp 062
Encompass_Preferred
Sulfate, Ion Calibration Standard 1000mg/L Martix H2O. This 1000 mg/L standard can be used for primary calibration or to prepare second source calibration check standards. It is traceable to NIST Standard Reference Materials, where available, and is guaranteed stable for one year.
Catalog No. 50-465-870