Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Certified Reference Material, Tin, 1000 mg/L, 125 mL

Catalog No. 50465860 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This 125 mL certified reference material (CRM) contains Tin at 1000 mg/L in an aqueous solution with hydrochloric acid. Designed for environmental calibration, it holds ISO 17034 accreditation, ensuring high quality and traceability.

  • Analyte: Tin
  • Concentration: 1000 mg/L
  • Volume: 125 mL
  • Matrix: H2O with HCl
  • Item type: Certified reference material
  • Accreditations: ISO 17034
  • Methods: EPA 200.7, EPA 200.8, EPA 282.1, EPA 282.2, EPA 6010, EPA 6010B, EPA 6020, EPA 7870, SM 3120
  • Storage: 20°C to 25°C (ambient)
  • Shelf life from date of manufacture: 36 months
  • Minimum shelf life on ship date: 12 months
  • Dangerous goods classification: Dangerous goods (DG)
  • Components per package: 1
  • Container type: 125 mL clear glass bottle
  • Formulation: Liquid
  • Application: Environmental

Catalog No. 50-465-860 Supplier Waters Corp Supplier No. 037
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This 125 mL certified reference material (CRM) contains Tin at 1000 mg/L in an aqueous solution with hydrochloric acid. Designed for environmental calibration, it holds ISO 17034 accreditation, ensuring high quality and traceability.

  • Analyte: Tin
  • Concentration: 1000 mg/L
  • Volume: 125 mL
  • Matrix: H2O with HCl
  • Item type: Certified reference material
  • Accreditations: ISO 17034
  • Methods: EPA 200.7, EPA 200.8, EPA 282.1, EPA 282.2, EPA 6010, EPA 6010B, EPA 6020, EPA 7870, SM 3120
  • Storage: 20°C to 25°C (ambient)
  • Shelf life from date of manufacture: 36 months
  • Minimum shelf life on ship date: 12 months
  • Dangerous goods classification: Dangerous goods (DG)
  • Components per package: 1
  • Container type: 125 mL clear glass bottle
  • Formulation: Liquid
  • Application: Environmental

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.