Learn More
Bellco Glass, Inc. 125mL Wide Mouth Bottle
SDP
Catalog No. 502329243
Encompass_Preferred
CELLTREAT Polypropylene Wide Mouth BottleSpecifications
Round shape and easypour wide mouth design Excellent chemical resistance and durable rigid walls Ideal for specimen and chemical storage Linerless leak proof screw top closure Assembled bottle and cap Size 125mL Pack Description 12/Bag Case Quantity 48
Catalog No. 50-232-9243 Supplier Bellco Glass, Inc. Supplier No. 229795000
CELLTREAT Polypropylene Wide Mouth BottleSpecifications
Round shape and easypour wide mouth design Excellent chemical resistance and durable rigid walls Ideal for specimen and chemical storage Linerless leak proof screw top closure Assembled bottle and cap Size 125mL Pack Description 12/Bag Case Quantity 48