Learn More
Ace Glass, Inc. 125mm powder funnel with 24/40 inner joint
SDP
Catalog No. 50974708
Encompass_Preferred
125mm powder funnel with 24/40 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-708 Supplier Ace Glass, Inc. Supplier No. 723611
125mm powder funnel with 24/40 inner joint is useful in pouring powders or liquids into ground joint containers.