Learn More
Ace Glass, Inc. 125mm powder funnel with 35/45
SDP
Catalog No. 50974709
Encompass_Preferred
125mm powder funnel with 35/45 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-709 Supplier Ace Glass, Inc. Supplier No. 723614
125mm powder funnel with 35/45 inner joint is useful in pouring powders or liquids into ground joint containers.