Learn More
Ace Glass, Inc. 190mm powder funnel with 45/50 inner joint
SDP
Catalog No. 50974714
Encompass_Preferred
190mm powder funnel with 45/50 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-714 Supplier Ace Glass, Inc. Supplier No. 7236145
190mm powder funnel with 45/50 inner joint is useful in pouring powders or liquids into ground joint containers.