Learn More
Ace Glass, Inc. 190mm powder funnel with 45/50 inner joint
SDP
Supplier:  Ace Glass, Inc. 723616
Encompass_Preferred
190mm powder funnel with 45/50 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-710