Learn More
Biotage 125MM SEP FLASK 19-22 TAPER
SDP
Catalog No. NC9850814
Encompass
125 mL Glass Separatory Funnel is a convenient way to collect the sample extract using the SPE-DEX 4790/3000/1000 Extractions systems. Perfect for Oil & Grease samples to easily drain the n-hexane and separate the residual water from the extraction solvent.
Catalog No. NC9850814 Supplier Biotage Supplier No. 27083401
125 mL Glass Separatory Funnel is a convenient way to collect the sample extract using the SPE-DEX 4790/3000/1000 Extractions systems. Perfect for Oil & Grease samples to easily drain the n-hexane and separate the residual water from the extraction solvent.