Learn More
Integra Biosciences Corp Sterile, Filter, Low Retention, 1250 ul Standard Tips, 5 VIAFLO 96 racks of 96 Tips
SDP
Catalog No. NC1341323
Encompass
Precise and consistent tip seal ensures that all tips on a MINI 96, VIAFLO 96 and VIAFLO 384 are at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.
Catalog No. NC1341323 Supplier Integra Biosciences Corp Supplier No. 6545
Precise and consistent tip seal ensures that all tips on a MINI 96, VIAFLO 96 and VIAFLO 384 are at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.