Learn More
Integra Biosciences Corp 125UL ECO RACKS OF 96 TIPS
SDP
Catalog No. NC2340469
Encompass
Sterile, FilterECO racks contain 60% less plastic than alternative racks, while maintaining the high standards that you expect from INTEGRA products.Once empty, ECO racks can be stacked and compressed to minimize the space needed in your lab's recycling, keeping your workspace clutter-free and supporting a positive working environment.
Catalog No. NC2340469 Supplier Integra Biosciences Corp Supplier No. 3728
Sterile, FilterECO racks contain 60% less plastic than alternative racks, while maintaining the high standards that you expect from INTEGRA products.Once empty, ECO racks can be stacked and compressed to minimize the space needed in your lab's recycling, keeping your workspace clutter-free and supporting a positive working environment.