Learn More
Integra Biosciences Corp 12.5 µl GRIPTIPS, Sterile, LONG, 5 XYZ Racks of 384 Tips
SDP
Supplier:  Integra Biosciences Corp 6405
Encompass
Precise and consistent tip seal ensures that all tips on MINI 96, VIAFLO 96 and VIAFLO 384 are at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate. For use with 2, 10, 12.5 and 20 µl INTEGRA pipettes. 44 mm length for easy access to 1.5 ml tubes
Catalog No. NC1279952