Learn More
AUTOMATION TRAINER 125 L, Clear, Filtered, Sterile, Racked (40-384-Tip Racks/Case)
SDP
Catalog No. NC2044894
Encompass_Preferred
Automation Trainer's Tecan MCA-Compatible 384-Format Pipette Tips are designed to be a direct replacement to Tecan's 384-Format 15 L, 50 L, & 125 L MCA tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required.
Catalog No. NC2044894 Supplier AUTOMATION TRAINER Supplier No. TM125CLFSR384
Automation Trainer's Tecan MCA-Compatible 384-Format Pipette Tips are designed to be a direct replacement to Tecan's 384-Format 15 L, 50 L, & 125 L MCA tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required.