Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Accuflow Systems Saint-Gobain C-Flex TPE Tubing, 0.125 in ID, 50 ft, Translucent

Catalog No. NC2312782
Encompass_Preferred

Translucent thermoplastic elastomer tubing engineered for biopharmaceutical and laboratory fluid transfer. The tubing is flexible, bondable, and suitable for peristaltic pumps, aseptic welding, and single-use assemblies; supplied as a 50 ft coil with a 0.125 in inner diameter and 0.25 in outer diameter.

  • Inner diameter 0.125 in (3.2 mm)
  • Outer diameter 0.250 in (6.4 mm)
  • Wall thickness 0.062 in (1.6 mm)
  • Standard length 50 ft (15 m)
  • Material thermoplastic elastomer (TPE)
  • Appearance translucent for fluid visibility
  • Temperature range -67°C to 135°C (-85°F to 275°F)
  • Average working pressure 27 psi (1.9 bar)
  • Sterilization compatible: gamma irradiation and X-Ray up to 50 kGy; autoclave one time at 121°C for 30 minutes (radiation recommended)
  • Applications peristaltic pumping, aseptic sealing and welding, single-use fluid transfer, buffer and media handling

Catalog No. NC2312782 Supplier Accuflow Systems Supplier No. 0821252
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Translucent thermoplastic elastomer tubing engineered for biopharmaceutical and laboratory fluid transfer. The tubing is flexible, bondable, and suitable for peristaltic pumps, aseptic welding, and single-use assemblies; supplied as a 50 ft coil with a 0.125 in inner diameter and 0.25 in outer diameter.

  • Inner diameter 0.125 in (3.2 mm)
  • Outer diameter 0.250 in (6.4 mm)
  • Wall thickness 0.062 in (1.6 mm)
  • Standard length 50 ft (15 m)
  • Material thermoplastic elastomer (TPE)
  • Appearance translucent for fluid visibility
  • Temperature range -67°C to 135°C (-85°F to 275°F)
  • Average working pressure 27 psi (1.9 bar)
  • Sterilization compatible: gamma irradiation and X-Ray up to 50 kGy; autoclave one time at 121°C for 30 minutes (radiation recommended)
  • Applications peristaltic pumping, aseptic sealing and welding, single-use fluid transfer, buffer and media handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.