Learn More
eMolecules​ 126-33-0 | Sulfolane | Oakwood Chemicals | MFCD00005484 | 120.170 | C4H8O2S | 99.000 | O=S1(=O)CCCC1 | 100g | 480131565
Catalog No. 5000158862 Shop All eMolecules Inc. Products
Encompass_Preferred
Sulfolane | Oakwood Chemicals | 126-33-0 | MFCD00005484 | 120.170 | C4H8O2S | 99.000 | O=S1(=O)CCCC1 | 100g | 480131565
Catalog No. 50-001-58862 Supplier eMolecules​ Supplier No. 480131565
Sulfolane | Oakwood Chemicals | 126-33-0 | MFCD00005484 | 120.170 | C4H8O2S | 99.000 | O=S1(=O)CCCC1 | 100g | 480131565