Learn More
Sino Biological Recombinant Human HE4/WFDC2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 12609H08H100
Encompass_Preferred
A DNA sequence encoding the human WFDC2 (NP_006094.3) (Met 1-Phe 124) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-8081