Learn More
Sino Biological Recombinant Human ELK1 Protein (His & GST Tag)
Supplier:  Sino Biological 12621H20B100
Encompass_Preferred
A DNA sequence encoding the human ELK1 (P19419-1) (Met 1-Pro 428) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8083