Learn More
Sino Biological Recombinant Human LAMTOR2 Protein (His Tag)
Supplier:  Sino Biological 12622H07E100
Encompass_Preferred
A DNA sequence encoding the mature form of human LAMTOR2 isoform 1 (Q9Y2Q5-1) (Met 1-Ser 125) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8085