Learn More
Sino Biological Recombinant Human S100P Protein
Supplier:  Sino Biological 12635HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human S100P (P25815) (Met 1-Lys 95) was expressed and purified.
Catalog No. 50-161-8089