Learn More
Sino Biological Recombinant Human S100P Protein, Amine-Labeled Biotinylated
Supplier:  Sino Biological 12635HNAEB20
Encompass_Preferred
A DNA sequence encoding the mature form of human S100P (P25815) (Met 1-Lys 95 ) was expressed and purified.
Catalog No. 50-161-8091