Learn More
Sino Biological Recombinant Human HOXA1 Protein (His Tag)
Supplier:  Sino Biological 12644H07E50
Encompass_Preferred
A DNA sequence encoding the mature form of human HOXA1 isoform 3 (P49639-1) (Met 1-His 335) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8095