Learn More
Sino Biological Recombinant Human SOD2/Mn-SOD Protein
Supplier:  Sino Biological 12656HNAE100
Encompass_Preferred
A DNA sequence encoding the mature form of human SOD2 (P04179-1) (Lys 25-Lys 222) was expressed and purified, with an initial Met.
Catalog No. 50-161-8098