Learn More
eMolecules​ 126705-22-4 | Fmoc-Acpc-OH | Combi-Blocks | MFCD00190873 | 323.348 | C19H17NO4 | 95.000 | OC(=O)C1(CC1)NC(=O)OCC1c2ccccc2-c2ccccc12 | 1g | 117566507
Catalog No. 5000162801 Shop All eMolecules Inc. Products
Encompass_Preferred
Fmoc-Acpc-OH | Combi-Blocks | 126705-22-4 | MFCD00190873 | 323.348 | C19H17NO4 | 95.000 | OC(=O)C1(CC1)NC(=O)OCC1c2ccccc2-c2ccccc12 | 1g | 117566507
Catalog No. 50-001-62801 Supplier eMolecules​ Supplier No. 117566507
Fmoc-Acpc-OH | Combi-Blocks | 126705-22-4 | MFCD00190873 | 323.348 | C19H17NO4 | 95.000 | OC(=O)C1(CC1)NC(=O)OCC1c2ccccc2-c2ccccc12 | 1g | 117566507