Learn More
Sino Biological Recombinant Human NEIL1 Protein (His Tag)
Supplier:  Sino Biological 12695H08E50
Encompass_Preferred
A DNA sequence encoding the human NEIL1 (AAH10876.1) (Met 1-Ser 390) was fused with a polyhistidine tag at the C-terminus and an initial Met at the N-terminus.
Catalog No. 50-161-8101