Learn More
eMolecules​ 1272758-17-4 | 2-Boc-2-azaspiro[4.5]decan-8-one | Accela ChemBio | MFCD18711592 | 253.342 | C14H23NO3 | 97.000 | CC(C)(C)OC(=O)N1CCC2(C1)CCC(=O)CC2 | 0.25g | 789348185
Catalog No. 5000489182 Shop All eMolecules Inc. Products
Encompass_Preferred
2-Boc-2-azaspiro[4.5]decan-8-one | Accela ChemBio | 1272758-17-4 | MFCD18711592 | 253.342 | C14H23NO3 | 97.000 | CC(C)(C)OC(=O)N1CCC2(C1)CCC(=O)CC2 | 0.25g | 789348185
Catalog No. 50-004-89182 Supplier eMolecules​ Supplier No. 789348185
2-Boc-2-azaspiro[4.5]decan-8-one | Accela ChemBio | 1272758-17-4 | MFCD18711592 | 253.342 | C14H23NO3 | 97.000 | CC(C)(C)OC(=O)N1CCC2(C1)CCC(=O)CC2 | 0.25g | 789348185