Learn More
Sino Biological Recombinant Human ISG15 Protein, HPLC-verified
Supplier:  Sino Biological 12729HNAE1
Encompass_Preferred
A DNA sequence encoding the Pro form of human ISG15 (AAH09507.1) (Met 1-Ser 165) was expressed and purified
Catalog No. 50-161-8116