Learn More
Sino Biological Recombinant Human ISG15 Protein (mature form), HPLC-verified
Supplier:  Sino Biological 12729HNAE11
Encompass_Preferred
A DNA sequence encoding the mature form of human ISG15 (AAH09507.1) (Met 1-Gly 157) was expressed and purified.
Catalog No. 50-161-8117