Learn More
Sino Biological Recombinant Human USP5 Protein (His Tag)
Supplier:  Sino Biological 12772H08B100
Encompass_Preferred
A DNA sequence encoding the human USP5 isoform short (P45974-2) (Met 1-Ser 835) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8124