Learn More
Sino Biological Recombinant Human MAD2L1 Protein (His Tag)
Supplier:  Sino Biological 12798H07E100
Encompass_Preferred
A DNA sequence encoding the human MAD2L1 (Q13257) (Met 1-Asp 205) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8127