Learn More
Sino Biological Recombinant Human Factor H-related 1/CFHR1 Protein (His Tag)
Supplier:  Sino Biological 12822H08H50
Encompass_Preferred
A DNA sequence encoding the human CFHR1 (Q03591) (Met1-Arg330) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8130