Learn More
Sino Biological Recombinant Human KIR2DL3 Protein (His Tag)
Supplier:  Sino Biological 12828H08H100
Encompass_Preferred
A DNA sequence encoding the human KIR2DL3 (AAX23102.1) extracellular domain (Met 1-His 245) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8131