Learn More
Sino Biological Recombinant Human SerpinB6 Protein (His Tag)
Supplier:  Sino Biological 12837H07B50
Encompass_Preferred
A DNA sequence encoding the human SERPINB6 (AAB30320.1) (Asp 2-Pro 376) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8134