Learn More
Sino Biological Recombinant Human SerpinB9 Protein (His Tag)
Supplier:  Sino Biological 12838H07B100
Encompass_Preferred
A DNA sequence encoding the human SERPINB9 (P50453) (Glu 2-Pro 376) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8135