Learn More
Sino Biological Recombinant Human ALDH4A1 Protein
Supplier:  Sino Biological 12856HNCB100
Encompass_Preferred
A DNA sequence encoding the human ALDH4A1 (AAH07581.1) (Lys 25-Gln 563) was expressed and purified with two additional amino acids (Gly & Pro ) at the N-terminus.
Catalog No. 50-161-8139