Learn More
Sino Biological Recombinant Human Ezrin Protein (His Tag)
Supplier:  Sino Biological 12858H07E50
Encompass_Preferred
A DNA sequence encoding the mature form of human EZR (P15311) (Pro2-Leu586) was expressed with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8142