Learn More
J&H Berge Inc 12880 uS/cm Conductivity Standard (500mL Bottle)
SDP
Catalog No. NC0850414
Encompass_Preferred
HI7030L is a premium quality 12,880 µS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution. Hanna conductivity solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna's line of conductivity standards have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle.
Catalog No. NC0850414 Supplier J&H Berge Inc Supplier No. HI7030L
HI7030L is a premium quality 12,880 µS/cm conductivity standard that is prepared against a NIST traceable potassium chloride solution. Hanna conductivity solutions have the lot number and expiration date clearly marked on the label and are air tight with a tamper-proof seal to ensure the quality of the solution. Hanna's line of conductivity standards have been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle.