Learn More
Sino Biological Recombinant Human MAX Protein (His & GST Tag)
Supplier:  Sino Biological 12885H20B50
Encompass_Preferred
A DNA sequence encoding the human MAX (NP_002373) (Met1-Ser160) was fused with the N-terminal polyhistidine-tagged GST tag at the N-terminus.
Catalog No. 50-161-8144