Learn More
Sino Biological Recombinant Human PTPMT1 Protein (His Tag)
Supplier:  Sino Biological 12902H07E50
Encompass_Preferred
A DNA sequence encoding the human PTPMT1 isoform 1 (Q8WUK0-1) (Lys 28-Thr 201) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8153