Learn More
Sino Biological Recombinant Human Claudin-11 Protein (mFc Tag)
Supplier:  Sino Biological 12903H04H100
Encompass_Preferred
A DNA sequence encoding the human CLDN11 (O75508) (Val23-Arg82) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-8154