Learn More
Sino Biological Recombinant Human UNG Protein (GST Tag)
Supplier:  Sino Biological 12939H09E100
Encompass_Preferred
A DNA sequence encoding the human UNG isoform 1 (P13051-2) (Phe 85-Leu 304) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-8158