Learn More
Sino Biological Recombinant Human PPM1A Protein (His Tag)
Supplier:  Sino Biological 12984H08E100
Encompass_Preferred
A DNA sequence encoding the human PPM1A isoform alpha-1 (P35813-1) (Met 1-Trp 382) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8164