Learn More
Sino Biological Recombinant Human SERPINB4 Protein (His Tag)
Supplier:  Sino Biological 12990H07B50
Encompass_Preferred
A DNA sequence encoding the human SERPINB4 (P48594) (Met 1-Pro 390) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-8167