Learn More
Sino Biological Recombinant Human EBP1 Protein (His Tag)
Supplier:  Sino Biological 12994H07E100
Encompass_Preferred
A DNA sequence encoding the human PA2G4 (Q9UQ80) (Ser 2-Asp 394) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8168