Learn More
USA Scientific Inc 12-cap strip for 0.2 mL, mixed
Catalog No. 502745479
Encompass
12-cap strip for 0.2 mL PCR tubes and plates, dome top, mixed colors, 80 strips (960 caps) per pack. Polypropylene.
Catalog No. 50-274-5479 Supplier USA Scientific Inc Supplier No. 14001208
12-cap strip for 0.2 mL PCR tubes and plates, dome top, mixed colors, 80 strips (960 caps) per pack. Polypropylene.